missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Glucose Oxidase Antibody Labeling Kit

Product Code. 18067945 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
300 μg
Unit Size:
1 set
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18067945 1 mg 1mg
18057375 300 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18067945 Supplier Novus Biologicals™ Supplier No. 7060015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

Glucose oxidase is an enzyme produced and purified from Aspergillus niger which catalyses the oxidation of glucose with the release of hydrogen peroxide. Its molecular weight is 80kDa, but it exists as a dimer of 160kDa Measurement of glucose by GOx is used in the food industry, in fermentation and, most importantly, as the basis of biosensors for the diagnosis of diabetes.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1 mg
Conjugate Gox
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-GOx Modifier reagent, 1 vial of LL-GOx Quencher reagent
Product Type Glucose Oxidase Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.