Learn More
Novus Biologicals™ Lightning-Link (R) Glucose Oxidase Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
30 μg
Unit Size:
1 set
Specifications
Specifications
| Description | Antibody Labeling Kit |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 30 μg |
| Target | Lightning-Link Glucose Oxidase |