missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Fluorescein Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
Fluorescein is a fluorophore widely used in biological analysis, with a pH dependent absorption and emission spectrum. In TBS LL-Fluorescein shows an absorption maximum at 494 nm and an emission maximum at 520 nm.
Order Info
Quantity
Specifications
Specifications
| Product Type | Fluorescein Antibody Labeling Kit |
| Content And Storage | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent. Store at -20°C. Avoid freeze-thaw cycles. |
| Conjugate | FITC |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?