missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) B-Phycoerythrin Antibody Labeling Kit

Product Code. 18778283 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
180 μg
60 μg
600 μg
Unit Size:
1 set
600µg
60µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778283 600 μg 600µg
18008565 60 μg 60µg
18758233 180 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18778283 Supplier Novus Biologicals™ Supplier No. 7160015
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Applications: Flow Cytometry

B-Phycoerythrin (B-PE) is a fluorescent protein from phycobiliprotein family. It has a primary absorption peak at 546 nm and a secondary peak at 563 nm. When excited at 546 nm it has a strong fluorescence emission with its maximum at 575 nm.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 600 μg
Conjugate B-Phycoerythrin
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-B-PE Modifier reagent, 1 vial of LL-B-PE Quencher reagent
Product Type B-Phycoerythrin Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.