Learn More
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit
Product Code. 18780304
Change view
Click to view available options
Quantity:
1 mg
1 x 100 μg Reaction
3 x 0.01 mg Reaction
300 μg
Unit Size:
1 set
100µg
1mg
30µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18780304 1 x 100 μg Reaction 100µg
18731204 1 mg 1mg
18736893 300 μg 1 set
18759733 3 x 0.01 mg Reaction 30µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18780304 Supplier Novus Biologicals™ Supplier No. 7650005
  • ONLINE EXCLUSIVE PRICE
    474.00€
    448.35€ / 100µg
    Save 25.65€
    5% Off
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Applications: Flow Cytometry
APC/Cy7 is a tandem conjugate. In the Lightning-Link APC/Cy7 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the APC in the form of long wavelength light at 776nm.
Order Info
Quantity
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1 x 100 μg Reaction
Conjugate APC/Cy7
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type APC-Cy7 Antibody Labeling Kit