Learn More
B Braun™ Lifosan™ Neutral Washing Lotion
Product Code. 11910777
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11910777 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11910777 Supplier B Braun™ Supplier No. 3864529
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
pH 5.5
Description Neutral Washing Lotion, 1L, 5.5 pH, for Normal to Oily Skin, Alkali and Soap Free, Protects Your Skin Against Irritation
For Use With (Application) Protects your skin against irritation
Quantity 1 Ea.
Volume (Metric) 1 L