Learn More
Afora™ Liebig Refrigerant
Product Code. 11965799
Change view
Click to view available options
Standard Taper Joint No:
14/23
Length (Metric):
120 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Unit Size
11965799 14/23 120 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11965799 Supplier Afora™ Supplier No. 1559/E
Estimated Shipment: 11-01-2027
Convert vapor refrigerant to liquid using accessories for the Afora™ Liebig Refrigerant Condenser, an important component in a refrigeration system.
  • Weight: 0.01kg
  • Liebig Refrigerant
Specifications
Description Liebig refrigerant
Length (Metric) 120 mm
Standard Taper Joint No 14/23