Learn More
BOLA Liebig Condensers Vacuum
Product Code. p-200109993
Click to view available options
Length (Metric) Jacket:
300 mm
450 mm
Unit Size:
Each
Product Code. 10144581
Brand: BOLA B29502
Only null left
Add to basket
Add to basket
Liebig Condensers Vacuum PTFE, with SAFE LAB-system
  • One-piece cooling tube with ground joint socket and cone size 29 made of PTFE, cooling jacket made of borosilicate glass with hose connectors made of PP and nuts for connection of cooling water.
  • The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Specifications
Certifications/Compliance FDA
Length (Metric) Jacket 300 mm