Learn More
BOLA Liebig Condensers Transparent
Click to view available options
Length (Metric) Jacket:
450 mm
600 mm
Unit Size:
Each
Description
- Thin-walled cooling tube made of translucent and gastight PFA
- cooling jacket made of borosilicate glass with hose connectors
- made of PP and nuts for connection of cooling water
- ground joint cone and socket size 29 made of PTFE.
- The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Length (Metric) Jacket | 450 mm |