Learn More
BOLA Liebig Condensers Transparent
Product Code. p-200109992
Click to view available options
Length (Metric) Jacket:
450 mm
600 mm
Unit Size:
Each
Product Code. 10247351
Brand: BOLA B29104
Only null left
Add to basket
Add to basket
Liebig Condensers Transparent PTFE, with SAFE LAB-system
  • Thin-walled cooling tube made of translucent and gastight PFA
  • cooling jacket made of borosilicate glass with hose connectors
  • made of PP and nuts for connection of cooling water
  • ground joint cone and socket size 29 made of PTFE.
  • The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Specifications
Certifications/Compliance FDA
Length (Metric) Jacket 450 mm