Learn More
Lenz Laborglasintrumente™ Liebig Condenser with GL Threads
Product Code. 10502503
Change view
Click to view available options
Standard Taper Joint No:
NS 29/32
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
10502503 NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10502503 Supplier Lenz Laborglasintrumente™ Supplier No. 05210314
  • ONLINE EXCLUSIVE PRICE
    62.85€
    47.24€ / Pack of 1
    Save 15.61€
    24% Off
Estimated Shipment: 21-12-2026
Liebig condenser for condensing vapours
  • Liebig condensers acc. to DIN 12576 and in addition to DIN
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Certifications/Compliance DIN 12576
Description Cooling Surface - 130 cm²
Length (Metric) Jacket 250 mm
Quantity 1
Standard Taper Joint No NS 29/32
Material DURAN™ Tubing