Learn More
Afora™ Liebig Condenser
Product Code. 12200103
Change view
Click to view available options
Standard Taper Joint No:
19/26
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No unitSize
12200103 19/26 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12200103 Supplier Afora™ Supplier No. 1563/E
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Creates condensate with large surface area of the straight inner tube which is surrounded by a larger straight outer tube. Afora™ Liebig Condenser is capable of condensing liquid from a much greater flow of incoming vapor. Considered the most efficient type of condenser.
  • Manufactured from blown glass
  • Cylindrical in shape
  • Inlet/outlet valves at opposite ends
  • Straight inner tube
  • Larger Straight outer tube
  • Complies with UNE 400316
Specifications
Description Surface exchange: 100cm2
Height (Metric) 361 mm
Length (Metric) Jacket 250 mm
Standard Taper Joint No 19/26
Material Blown Glass