Learn More
Haldenwanger™ Lids for Porcelain Crucibles
Product Code. 12356507
Change view
Click to view available options
Diameter (Metric) Outer:
28mm
32mm
35mm
52mm
60mm
68mm
Unit Size:
Each
Pack of 10
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
12356507 35mm Pack of 10
12867713 68mm Pack of 5
12346507 28mm Pack of 10
12376487 32mm Pack of 10
12306497 60mm Pack of 5
12366507 52mm Pack of 10
10353121 52mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12356507 Supplier Haldenwanger™ Supplier No. L 79D/7
  • ONLINE EXCLUSIVE PRICE
    73.35€
    69.58€ / Pack of 10
    Save 3.77€
    5% Off
Estimated Shipment: 09-10-2026
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Diameter (Metric) Outer 35mm
Product Type Crucible Lid