Learn More
Haldenwanger™ Lids For Porcelain Crucibles
Product Code. 12396487
Click to view available options
For Use With (Application):
13mL crucibles
20mL crucibles
Unit Size:
Pack of 10
Product Code. 12396487
Brand: Haldenwanger™ L 79D/6
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed to fit Haldenwanger™ Porcelain Crucibles.
  • Available in a variety of sizes
  • For use with porcelain crucibles
Specifications
Color White
Material Porcelain
For Use With (Application) 20mL crucibles
Diameter (Metric) Outer 46mm
Product Type Crucible Lids