missing translation for 'onlineSavingsMsg'
Learn More

LGK 974, TRC

Product Code. 30468877
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468877 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468877 Supplier Toronto Research Chemicals Supplier No. TRCL39764050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material LGK 974
CAS 1243244-14-5
Molecular Formula C23 H20 N6 O
Quantity 50 mg
Synonym 2',3-Dimethyl-N-[5-(2-pyrazinyl)-2-pyridinyl]-[2,4'-bipyridine]-5-acetamide, 2-(2',3-Dimethyl-[2,4'-bipyridin]-5-yl)-N-(5-(pyrazin-2-yl)pyridin-2-yl)acetamide
InChI Formula InChI=1S/C23H20N6O/c1-15-9-17(12-28-23(15)18-5-6-25-16(2)10-18)11-22(30)29-21-4-3-19(13-27-21)20-14-24-7-8-26-20/h3-10,12-14H,11H2,1-2H3,(H,27,29,30)
SMILES Cc1cc(ccn1)c2ncc(CC(=O)Nc3ccc(cn3)c4cnccn4)cc2C
IUPAC Name 2-[5-methyl-6-(2-methylpyridin-4-yl)pyridin-3-yl]-N-(5-pyrazin-2-ylpyridin-2-yl)acetamide
Molecular Weight (g/mol) 396.44
Formula Weight 396.1699
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.