missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A blocking peptide containing 12 amino acids near the center of human LGI3. Source: Synthetic (Accession #: AAQ88483) The Lgi3 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 203190 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-76383. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | Leucine-rich glioma-inactivated protein 3, leucine-rich repeat LGI family member 3, leucine-rich repeat LGI family, member 3, LGI1-like protein 4, LGIL4 |
| Gene Symbol | LGI3 |
| Product Type | Lgi3 |
| Quantity | 0.05 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?