missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LG 100268

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
LG 100268 is a potent and selective retinoid X receptor (RXR) agonist (EC50 = 4 nM). Displays >1000-fold selectivity for RXR over RAR. Activates RXR homodimers to induce transcriptional activation. Induces tissue transglutaminase (TGase) activity in HL-60 human promyelocytic leukemia cell line. Also activates RXR/PPARγ heterodimers.
Specifications
Specifications
| Chemical Name or Material | 6-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)cyclopropyl]-3-pyridinecarboxylic acid |
| CAS | 153559-76-3 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C24H29NO2 |
| Formula Weight | Observed MW: 363.49 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction