missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levulinic-3,3,5,5,5-d5 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Levulinic-d5 Acid |
| CAS | 1206185-52-5 |
| Molecular Formula | C5 D5 H3 O3 |
| Quantity | 0.05 g |
| Synonym | 3-Acetylpropionic-d5 Acid, 3-Oxobutanecarboxylic-d5 Acid, 4-Ketovaleric-d5 Acid, 4-Oxopentanoic-d5 Acid, 4-Oxovaleric-d5 Acid, Laevulinic-d5 Acid, Levulic-d5 Acid, NSC 3716, β-Acetylpropionic-d5 Acid, γ-Ketovaleric-d5 Acid, Levulinic-3,3,5,5,5-d5 Acid |
| InChI Formula | InChI=1 S/C5H8O3/c1-4(6)2-3-5(7)8/h2-3H2,1H3,(H,7,8)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])CC(=O)O |
| IUPAC Name | 3,3,5,5,5-pentadeuterio-4-oxopentanoic acid |
| Molecular Weight (g/mol) | 121.15 |
| Formula Weight | 121.0787 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction