missing translation for 'onlineSavingsMsg'
Learn More

Leuprolide Acetate EP Impurity H, TRC

Product Code. 30004756
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004756 5 mg 5mg
30138291 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004756 Supplier Toronto Research Chemicals Supplier No. TRCL3306355MG
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112710-58-4
Synonym Luteinizing hormone-releasing factor (swine), 6-D-leucine-7-D-leucine-9-(N-ethyl-L-prolinamide)-10-deglycinamide- (9CI)∣Luteinizing hormone-releasing factor (pig), 6-D-leucine-7-D-leucine-9-(N-ethyl-L-prolinamide)-10-deglycinamide-∣Leuprolide Acetate EP Impurity H

Specifications

Format Neat
Molecular Weight (g/mol) 1209.4
Quantity 5 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.