Learn More
Leupeptin, MP Biomedicals™
Product Code. 11448191 Shop All MP Biomedical Products
Click to view available options
Quantity:
5mg
25mg
100mg
Unit Size:
100mg
25mg
5mg
Product Code. 11448191
Brand: MP Biomedicals 0215155325
Inhibits trypsin-like proteases and some cysteine proteases. MP Biomedicals' Leupeptin is a modified tripeptide reversible protease inhibitor. Little to no inhibition is seen against pepsin, cathepsins A and D, and alpha-chymotrypsin.
When adjusted for molarity, all three salt forms are equally effective; however, the hydrochloride salt is usually less invasive in biological settings.
Inhibits trypsin-like proteases and some cysteine proteases including: endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.
TRUSTED_SUSTAINABILITY
Specifications
CAS 103476-89-7
Content And Storage Store at −20°C, desiccate, protect from light.
Format White Powder
Molecular Weight (g/mol) 427.57
Quantity 25mg