missing translation for 'onlineSavingsMsg'
Learn More

Melford Leupeptin Hemisulfate

Product Code. 30018380
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 30018380

Brand: Melford L220350.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.

Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin. Working concentration: 1-10uM

Specifications

CAS 103476-89-7
Molecular Formula C20H38N6O4 1/2H2SO4
Quantity 10 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.