Learn More
BD Biosciences™ Leucocount™ Kit (CE/IVD)
Product Code. 15855908
Click to view available options
Quantity:
50 Tests
Unit Size:
1 set
Product Code. 15855908
Brand: BD Biosciences™ 340523
Count residual white blood cells (rWBCs) in leucoreduced blood products.
  • Designed for counting residual white blood cells (rWBCs) in leucoreduced blood products.
  • This product is CE marked to the IVD directive 98/79/EC and available in Europe for use on the BD FACSVia™ system and the BD FACSCalibur™ flow cytometer.
  • Contents: BD Leucocount™ reagent with BD Trucount™ tubes.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage PI Nucleic Acid Dye, 0.1% Sodium Azide, RNAse, Detergent, Buffers
Includes Trucount™ Tubes
No. of Tests 50 Tests
Quantity 50 Tests
Product Type Enumeration Kit