Learn More
BD Biosciences™ Leucocount™ Kit (CE/IVD)
Click to view available options
Quantity:
50 Tests
Unit Size:
1 set
Description
- Designed for counting residual white blood cells (rWBCs) in leucoreduced blood products.
- This product is CE marked to the IVD directive 98/79/EC and available in Europe for use on the BD FACSVia™ system and the BD FACSCalibur™ flow cytometer.
- Contents: BD Leucocount™ reagent with BD Trucount™ tubes.
Specifications
Specifications
| Content And Storage | PI Nucleic Acid Dye, 0.1% Sodium Azide, RNAse, Detergent, Buffers |
| Includes | Trucount™ Tubes |
| No. of Tests | 50 Tests |
| Quantity | 50 Tests |
| Product Type | Enumeration Kit |