Learn More
Thermo Scientific Chemicals [Leu8] Renin Substrate Tetradecapeptide, Rat
Click to view available options
Substrate Quantity:
1mg
5mg
Unit Size:
1mg
5mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to off-white |
| Form | Solid |
| Molecular Weight (g/mol) | 1789.06 |
| Storage Requirements | -30° to -10°C |
| Substrate Quantity | 1mg |