Learn More
LEPRE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553527
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: INKLEKAVAA AHTFFVGNPE HMEMQQNLDY YQTMSGVKEA DFKDLETQPH MQEFRLGVRL YSEEQPQEAV PHLEAALQEY FVAYEECRAL CEGPYDYDGY NYLEYNADLF QAITDHYIQV LNCKQNCVTE LASHP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
Basement membrane-associated chondroitin sulfate proteoglycan (CSPG). Has prolyl 3-hydroxylase activity catalyzing the post-translational formation of 3-hydroxyproline in -Xaa-Pro- Gly- sequences in collagens, especially types IV and V. May be involved in the secretory pathway of cells. Has growth suppressive activity in fibroblasts.Specifications
LEPRE1 | |
Polyclonal | |
Unconjugated | |
P3H1 | |
GROS1; Growth suppressor 1; LEPRE1; leprecan; Leprecan-1; leucine- and proline-enriched proteoglycan 1; leucine proline-enriched proteoglycan (leprecan) 1; OI8; P3H1; procollagen-proline 3-dioxygenase; prolyl 3-hydroxylase 1; PSEC0109 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q32P28 | |
P3H1 | |
Recombinant protein corresponding to Human LEPRE1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only