Learn More
Emma Safety Footwear Leo Safety Shoes
Click to view available options
Size:
35
36
37
38
39
48
49
missing translation for 'unitSize'
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather
- GripForce™ Easy Twist
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Laces |
| Color | Black |
| Design | Leo |
| Liner Material | Hydro-Tec™ Sanitized Silver |
| Material | Leather |
| Size | 39 |
| Toe Material | Steel |