Learn More
Emma Safety Footwear LeMans Optimal Ventilation XD-Fit Shoes
Product Code. 10307274
Change view
Click to view available options
Size:
40
41
Unit Size:
1 pair
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10307274 40 1 pair
10032424 41 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10307274 Supplier Emma Safety Footwear Supplier No. MM72756040
  • ONLINE EXCLUSIVE PRICE
    163.26€
    105.88€ / 1 pairs
    Save 57.38€
    35% Off
Estimated Shipment: 23-12-2026
Designed to provide perspiration and moisture removal.
  • Nubuck uppers
  • Hydro-Tec™ sanitized silver lining
  • Non-woven antistatic insole
  • GripForce™ Easy Twist outsole
Specifications
Color Black, Gray
Liner Material Hydro-Tec sanitized silver lining
Size 40
Sole Features Non-woven antistatic insole
Sole Material GripForce™ Easy Twist outsole
Product Type Optimal Ventilation Shoe