Learn More
Novus Biologicals™ Lectin Kit I Fluorescein Kit
Product Code. 18352168 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352168 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18352168 Supplier Novus Biologicals™ Supplier No. FLK2100NB
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
For Use With (Application) Immunofluorescence
Quantity 1 Kit
Storage Requirements Store at 4°C.
Protein Lectin Kit I Fluorescein