missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Pb/Sn/Ca Battery Alloy, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Pb/Sn/Ca Battery Alloy, 50g
Specifications
Specifications
| Chemical Name or Material | Mercury/Magnesium/Arsenic/Indium/Sulfur/Tellurium/Nickel/Copper/Antimony/Cadmium/Silver/Bismuth/Zinc/Aluminium/Calcium/Tin/Lead |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Mercury -70 ppm/Magnesium -30 ppm/Arsenic -20 ppm/Indium -10 ppm/Sulfur -10 ppm/Tellurium -4 ppm/Nickel -1 ppm/Copper 0.0014 %/Antimony 0.0017 %/Cadmium 0.0058 %/Silver 0.0061 %/Bismuth 0.0089 %/Zinc 0.024 %/Aluminium 0.025 %/Calcium 0.078 %/Tin 0.515 %/Lead 99.31 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction