Learn More
5-Amino-1-phenylpyrazole-4-carboxamide, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 5-Amino-1-phenylpyrazole-4-carboxamide |
| CAS | 50427-77-5 |
| Molecular Formula | C10 H10 N4 O |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C10H10N4O/c11-9-8(10(12)15)6-13-14(9)7-4-2-1-3-5-7/h1-6H,11H2,(H2,12,15) |
| SMILES | NC(=O)c1cnn(c1N)c2ccccc2 |
| IUPAC Name | 5-amino-1-phenylpyrazole-4-carboxamide |
| Molecular Weight (g/mol) | 202.21 |
| Formula Weight | 202.0855 |