Learn More
Lead(Ii) Oxide, ReagentPlus™, ≥99.9% trace metals basis, Honeywell™
Product Code. p-7120510 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Product Code. 15641940
Brand: Honeywell Research Chemicals 211907100G
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Powder, <10 μm, Reagent Grade,≥99.9% trace metals basis
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
IUPAC Name oxolead
SMILES O=[Pb]
Specifications
Boiling Point 1470°C
Melting Point 890°C
Color Yellow
Physical Form Powder
Quantity 100 g
Linear Formula PbO
UN Number UN2291
Formula Weight 223.2g/mol
Percent Purity ≥99.9% (trace metals basis)
Grade ReagentPlus™
Packaging Poly Bottle
Vapor Pressure 10mmHg (0°C)
Density 9.66g/cu.cm
Chemical Name or Material Lead(Ii) Oxide