Learn More
Lead(II) acetate, basic
Product Code. 11452778
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
Product Code. 11452778
Brand: Thermo Scientific Alfa Aesar A16763.0E
Request Bulk or Custom Format
CAS: 1335-32-6 | C4H6O4Pb·2Pb(OH)2
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1335-32-6
Molecular Formula C4H6O4Pb·2Pb(OH)2
MDL Number MFCD00013041
Synonym Lead subacetate
Specifications
Linear Formula Pb(OOCCH3)2·2Pb(OH)2
Quantity 2500 g
UN Number UN1616
Sensitivity Air sensitive
Merck Index 14,5419
Formula Weight 807.72
Chemical Name or Material Lead(II) acetate, basic
compliance-icons compliance-icons
Product Identifier
  • Lead(II) acetate, basic
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .