missing translation for 'onlineSavingsMsg'
Learn More

LDS-sample buffer (4X), reducing, Thermo Scientific Chemicals

POA - 45.11€


Products 2
Product Code Brand Price Quantity & Availability  
Product Code Brand Price Quantity & Availability  
15454419
SDS missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
J61942.AD
POA
50mL
View alternative products
missing translation for 'toSaveUpto' 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15444419
SDS missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
J61942.AC
25mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description

Description

LDS-sample buffer (4X), reducing is used to prepare protein samples for denaturing gel electrophoresis. It is used in 1D gel electrophoresis, bis-tris gel electrophoresis, tris-acetate gel electrophoresis, protein Gel electrophoresis, proteins, expression, isolation & analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
LDS-sample buffer (4X), reducing is used to prepare protein samples for denaturing gel electrophoresis. It is used in 1D gel electrophoresis, bis-tris gel electrophoresis, tris-acetate gel electrophoresis, protein Gel electrophoresis, proteins, expression, isolation & analysis.

Solubility
Not miscible in water.

Notes
This buffer contains 988mM Tris-HCl, 2.04mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue, and 0.7mM phenol red, and 8% mercaptoethanol at pH 7.5.
Videos
SDS
Documents

Documents

Certificates

RUO – Research Use Only