missing translation for 'onlineSavingsMsg'
Learn More

LDS-sample buffer (4X), reducing, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar J61942.AD

Additional Details : Weight : 0.00050kg Transport : UN number : 3082 Chem class : 9 Pack group : III


 View more versions of this product

Product Code. 15454419

  • POA / 50mL
Estimated Shipment: 28-05-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

LDS-sample buffer (4X), reducing is used to prepare protein samples for denaturing gel electrophoresis. It is used in 1D gel electrophoresis, bis-tris gel electrophoresis, tris-acetate gel electrophoresis, protein Gel electrophoresis, proteins, expression, isolation & analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
LDS-sample buffer (4X), reducing is used to prepare protein samples for denaturing gel electrophoresis. It is used in 1D gel electrophoresis, bis-tris gel electrophoresis, tris-acetate gel electrophoresis, protein Gel electrophoresis, proteins, expression, isolation & analysis.

Solubility
Not miscible in water.

Notes
This buffer contains 988mM Tris-HCl, 2.04mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue, and 0.7mM phenol red, and 8% mercaptoethanol at pH 7.5.
Specifications

Specifications

Blue
50 mL
Viscous Liquid
4X
Ambient Temperatures
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • LDS-sample buffer (4X), reducing

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid, monododecyl ester, lithium salt, 2-Mercaptoethanol, Phenol, 4,4'-(3H-2,1-benzoxathiol

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only