Learn More
Kautex™ LDPE Screw Closure
Product Code. p-8013593
Click to view available options
Diameter (Metric):
18 mm
25 mm
28 mm
40 mm
50 mm
65 mm
Unit Size:
Each
Pack of 100
Product Code. 11950867
Brand: Kautex™ 2000070604
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Feature LDPE light weight, tough construction. Kautex™ LDPE Screw Closures are available in a wide variety of sizes for Kautex™ narrow and wide neck bottles.
  • Made from low density polyethylene
  • Translucent
  • Available for a wide variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material LDPE
Thread Size (Metric) 00 mm
Neck Finish 00 mm
For Use With (Equipment) 50/100/200 mL Narrow Neck Bottle
Diameter (Metric) 18 mm