Learn More
Buerkle™ LDPE, Mixing/Dosing Container
Product Code. p-7230556 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
60 L
100 L
200 L
500 L
Unit Size:
Each
Product Code. 10086641
Brand: Buerkle™ 34600060
Only null left
Add to basket
Add to basket
The containers close completely tight. They have thick sides, they are seamless, impact resistant and stressfree sintered. The wide-necked opening enables safe filling.
  • Containers close completely tight
  • Seamless, thick sides
  • Impact-resistant and stress-free sintered
  • Manufactured from LDPE
  • With 0.75 in. inner thread at the container base
  • Suitable for foodstuffs
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 60 L
Closure Color Blue
Closure Material LDPE
Closure Size 0.75 in.
Closure Type Screw Cap
Color Translucent
Diameter (Metric) Outer 450 mm
Diameter (Metric) Outer Neck 150 mm
For Use With (Application) Foodstuffs
Height (Metric) 600 mm
Material LDPE
Mouth Style Wide
No. per Pack 1 Ea.
Product Type Mixing/Dosing Container
Shape Round