Learn More
Buerkle™ LDPE Rectangular Container
Product Code. p-8011242 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
23 L
45 L
48 L
50 L
150 L
155 L
Unit Size:
Each
Depending on temperature and the expected contents, the rectangular containers may have to be strengthened with a frame. Kindly you ask us.
Product Code. 10106310
Brand: Buerkle™ 34400235
Depending on temperature and the expected contents, the rectangular containers may have to be strengthened with a frame. Kindly you ask us.
LDPE Rectangular Container for various applications. Produced seamlessly and stress-relieved with the LaboPlast™ sintering method. The wall thickness increases downwardly. This guarantees optimum ratio of wall thickness to filling load.
  • L-LDPE (linear LDPE), transparent
  • Rupture-proof
TRUSTED_SUSTAINABILITY
Specifications
Product Type Container
Material LDPE
Shape Rectangular
Capacity (Metric) 23 L
For Use With (Application) Suitable For Use With Foodstuff
Height (Metric) 410 mm
Length (Metric) 220 mm
No. per Pack 1 Ea.
Width (Metric) 220 mm