missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LDN 193188

Description
LDN 193188 is an inhibitor of phosphatidylcholine transfer protein (PC-TP). Regulates hepatic glucose metabolism. Also promotes insulin-independent phosphorylation of key insulin signaling molecules.
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-N-[[[4-[[(4,6-dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]amino]carbonyl]benzamide |
| CAS | 1267610-30-9 |
| Quantity | 10 mg |
| Target | Lipid Metabolism Inhibitors |
| Molecular Formula | C20H17Cl2N5O4S |
| Formula Weight | 494.35 |
| Purity | >99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?