missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LDN 193188
GREENER_CHOICE

Product Code. 18714958
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18714958 50 mg 50mg
18731568 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 18714958

Brand: Tocris Bioscience™ 5631/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of phosphatidylcholine transfer protein (PC-TP)

LDN 193188 is an inhibitor of phosphatidylcholine transfer protein (PC-TP). Regulates hepatic glucose metabolism. Also promotes insulin-independent phosphorylation of key insulin signaling molecules.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichloro-N-[[[4-[[(4,6-dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]amino]carbonyl]benzamide
CAS 1267610-30-9
Quantity 50 mg
Target Lipid Metabolism Inhibitors
Molecular Formula C20H17Cl2N5O4S
Formula Weight 494.35
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.