Learn More
Taylor Wharton™ LD Series Cryogenic Storage Dewar
Product Code. p-8015010
Click to view available options
Capacity (Metric):
5 L
10 L
50 L
Unit Size:
Each
Product Code. 11768519
Brand: Taylor Wharton™ LD10
Only null left
Add to basket
Add to basket
The LD series of cryogenic dewars are used to store and distribute small amounts of liquid nitrogen. They feature extremely low evaporation rates and are convenient to use for various applications.
Specifications
Type Cryogenic Storage Dewar
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Height (English) Exterior 23.5 in.
Height (Metric) Exterior 597 mm
Weight (English) 14.6 lb.
Weight (Metric) 6.6 kg