missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LCL 161, phenol
GREENER_CHOICE

Product Code. 18768556
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18768556

Brand: Tocris Bioscience™ 7178/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Functionalized IAP ligand for PROTACs

LCL 161, phenol is a functionalized IAP (XIAP and cIAP) ligand for PROTAC™ research and development. Boc protected LCL 161. Supplied with a phenol functional handle for ready conjugation to a linker/target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl ((S)-1-(((S)-1-cyclohexyl-2-((S)-2-(4-(3-hydroxybenzoyl)thiazol-2-yl)pyrrolidin-1-yl)-2-oxoethyl)amino)-1-oxopropan-2-yl)(methyl)carbamate
CAS 2095244-42-9
Quantity 50 mg
Target Degrader Building Blocks
Molecular Formula C31H42N4O6S
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.