Learn More
MP Biomedicals™ LB Medium, Dehydrated (Capsules)
Product Code. 11303229 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
227 g
454 g
1000 g
5000 g
Unit Size:
1kg
227g
454g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11303229 454 g 454g
11373229 5000 g 5kg
11343229 1000 g 1kg
11373219 227 g 227g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11303229 Supplier MP Biomedicals™ Supplier No. 113002021
  • ONLINE EXCLUSIVE PRICE
    170.00€
    161.70€ / 454g
    Save 8.30€
    4% Off
 Restricted Item
In Stock
Formulations for the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter, and LB Medium contains 10g NaCl per liter
Specifications
Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Form Dehydrated
Product Type Microbiology Media
Formulation Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L
For Use With (Application) For growth of Escherichia coli, plasmid and other bacterial cultures in liquid medium
Quantity 454 g