Learn More
Fisher BioReagents™ LB Broth, Lennox (Pre-Buffered Capsules)
Click to view available options
Quantity:
25 x 20 g
500 x 1 g
Packaging Type:
Poly Bottle
Unit Size:
500g
Beskrivning
- Directions for Media Preparation: Add 1 capsule per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
- Product is in capsule form
- Each capsule contains 20g of broth
- Capsules are prebuffered and set to pH 7.2 at 20°C using a 10mM solution of tris/tris HCl
- pH will vary with temperature
Spezifikation
Spezifikation
| Format | Pre-Buffered Capsules |
| Media Type | LB Broth, Lennox |
| Product Type | Microbiology Media |
| Formulation | Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L, TRIS/TRIS HCl 1.5g/L |
| For Use With (Application) | For maintenance and propagation of Escherichia coli in molecular biology procedures |
| Packaging Type | Poly Bottle |
| Quantity | 25 x 20 g |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur