missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LB Agar with Ampicillin
Description
Support bacterial growth while inhibiting resistant strains with LB Agar enriched with Ampicillin, providing reliable results.
Specifications
Specifications
| Color | Transparent |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm |
| Manufacturing Origin | Germany |
| Product Type | Agar Plate |
| Fill Volume | 17 mL |
| Quantity | 10 Plates |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?