Learn More
Novus Biologicals™ Latex and Europium Conjugated Antibody Purification Kit
Product Code. 18176090 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
4 Units
Unit Size:
4 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18176090 4 Units 4 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18176090 Supplier Novus Biologicals™ Supplier No. 10200040
Easy-to-use kit for the removal of interfering buffer components that can affect the conjugation reaction when using LATEX™ Conjugation Kits.
The kit utilizes a simple spin column to clean-up the antibody by buffer exchanging to remove the unwanted buffer components. The kit can also be used for concentrating your antibody by recovering the antibody in a smaller volume than the volume initially added to the Spin Cartridge.
Kit Contents:
  • 1 vial 20x Reaction buffer A
  • 1 vial 20x Reaction buffer B
  • 4 spin cartridge / collecting tube assemblies
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C
Quantity 4 Units
Target LATEX™ and Europium Conjugated