Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Staphylococcus aureus
Product Code. 15988012 Shop All Novacyt Group Products
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
100 tests
50 tests
500 tests
Product Code. 15988012
Brand: Novacyt - Microgen Bioproducts M43CE
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Can be used to obtain rapid, presumptive-positive results from selective media for targeted identification/diagnosis.
  • Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
  • Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
  • Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
Content And Storage 2°C to 8°C
Detectable Analytes S. aureus
Type Latex Agglutination Test
Quantity 100 Tests
Test Time 2 min.