Learn More
BRAND™ Lateral Burette Stopcock for Burette, Amber
Shop All Brand Products
Click to view available options
For Use With (Equipment):
25 mL Burette
25 mL and 50 mL Burette
50 mL Burette
Unit Size:
Each
Description
- Stopcock housing with precision Borosilicate Glass 3.3 burette tip
- Interchangeable borosilicate glass key with retention device
- Stopcock size 3NS/12
Specifications
Specifications
| Type | Burette Stopcock |
| Material | Amber Borosilicate Glass (3.3) |
| For Use With (Equipment) | 25 mL and 50 mL Burette |
| Includes | Glass Key |