Learn More
KGW Isotherm™ Large Insulating Dewar Flasks, Blue with Lid and Grips
Click to view available options
Capacity (Metric):
28 L
Unit Size:
Each
Description
- Easy handling with two side grips
- Insulated lid made of white-coated aluminum with plastic grip
- Made of fully silvered borosilicate glass according to ISO 3585
- Protective casing made of blue-coated metal
- Area of application:
- Flexible laboratory, medical and biotechnology applications
- Store/transport liquid nitrogen (N2), dry ice (CO2) and temperature sensitive goods
- Suitable as an insulating flask
- Chemical characteristics:
- Hydrolytic resistance: according to ISO 719 (98°C)
- Hydrolytic resistance: according to ISO 720 (121°C)
- Acid resistance: according to ISO 1776
- Alkaline resistance: according to ISO 695-A2
- Temperature range:
- Glass refill from -200°C to + 150°C, for a short time 200°C
- Casing with mounting rubber and Latizell buffer from -10°C to + 60°C
Specifications
Specifications
| Cap Lid Type | Insulated, White coated aluminum, loose lied, with handle |
| Color | Blue |
| Diameter (Metric) Outer | 28-30 cm |
| For Use With (Application) | Storage and transport of temperature sensitive goods e.g. dry ice |
| Height (Metric) Inner | 62 cm |
| Height (Metric) | 75 cm |
| Includes | Side grips, Lid |
| Model | 34 C |
| Base Material | Metal |
| Capacity (Metric) | 28 L |
| Show More |