Learn More
Lanthanum(III) Trifluoromethanesulfonate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Lanthanum(III) Trifluoromethanesulfonate |
| CAS | 52093-26-2 |
| Molecular Formula | C3F9LaO9S3 |
| Quantity | 1 g |
| Synonym | 1,1,1-Trifluoromethanesulfonic Acid Lanthanum(3+) Salt, Trifluoromethanesulfonic Acid Lanthanum(3+) Salt, Lanthanum Triflate, Lanthanum Trifluoromethanesulfonate, Lanthanum Tris(trifluoromethanesulfonate), Lanthanum(3+) Triflate, Lanthanum(3+) Trifluoromethanesulfonate, Lanthanum(III) Triflate, Tris(trifluoromethanesulfonato)lanthanum |
| InChI Formula | InChI=1S/3CHF3O3S.La/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3 |
| SMILES | [O-]S(C(F)(F)F)(=O)=O.[O-]S(C(F)(F)F)(=O)=O.[O-]S(C(F)(F)F)(=O)=O.[La+3] |
| IUPAC Name | lanthanum(III) trifluoromethanesulfonate |
| Molecular Weight (g/mol) | 586.11 |
| Formula Weight | 586.76 |