Learn More
Brady™ LAN Network Card, Ethernet
Description
- Accessory for the BMP™51/53, BBP™33 and BBP™85 Printers.
Specifications
Specifications
| Product Type | Network Card |
| Color | Gray |
| For Use With (Equipment) | BBP31 Sign and Label Printer, BBP33 Label Printer, BBP85 Industrial Sign and Label Printer, BMP51 Label Printer, BMP53 Label Printer |
| Description | LAN Network Card |
| Format | Label Maker Attachment |
| Quantity | 1 Ea. |
| Alternate Part Number | NET-LAN |