Learn More
UVP Lamp XX-40BLB bench brackets included aluminum 220-240V 50/60Hz length 1260mm supplied with 2 x 365nm BLB 40W tubes UVP
Product Code. 12832979
Change view
Click to view available options
Voltage:
220/240 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
12832979 220/240 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12832979 Supplier UVP Supplier No. 95004305