Learn More
Brady™ Laminated Polyester Warning Sign
Product Code. 11720194
Change view
Click to view available options
Height (Metric):
50 mm
100 mm
200 mm
315 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
11720194 315 mm Each
10000674 100 mm Each
10481834 200 mm Each
10653385 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11720194 Supplier Brady™ Supplier No. 223438
Only null left
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC 300-TRI 315-B7541
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipemarkers and safety signs
Height (Metric) 315 mm
Thickness 0.024 mm
Width (Metric) 315 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Warning of Flammable Materials
Product Type Warning Sign
Color Black on Yellow